missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid, 97%

Catalog No. AAH52569MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52569MD 250 mg
AAH5256903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52569MD Supplier Thermo Scientific Chemicals Supplier No. H52569MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 73397-27-0 | C12H23NO5 | 261.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73397-27-0
Molecular Formula C12H23NO5
Molecular Weight (g/mol) 261.32
MDL Number MFCD04974450
InChI Key DJZCWTDKDFJARG-UHFFFAOYNA-N
Synonym 2s,3s-3-tert-butoxycarbonyl amino-2-hydroxy-5-methylhexanoic acid, n-boc-2s,3s-2-hydroxy-3-amino-5-methylhexanoic acid, 2s,3s-2-hydroxy-5-methyl-3-2-methylpropan-2-yl oxycarbonylamino hexanoic acid, 2s,3s-3-t-butoxycarbonyl amino-2-hydroxy-5-methylhexaneoic acid, 2s,3s-2-hydroxy-3-tert-butyloxycarbonyl amino-5-methylhexanoic acid, 2s,3s-3-tert-butoxycarbonyl amino-2-hydroxy-5-methylhexanoicacid, hexanoic acid,3-1,1-dimethylethoxy carbonyl amino-2-hydroxy-5-methyl-, 2s,3s
PubChem CID 2762291
SMILES CC(C)CC(NC(=O)OC(C)(C)C)C(O)C(O)=O

Specifications

Quantity 250 mg
IUPAC Name 3-{[(tert-butoxy)carbonyl]amino}-2-hydroxy-5-methylhexanoic acid
Formula Weight 261.32
Percent Purity 97%
Chemical Name or Material (2S,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only