missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-2-Chloro-3-methylvaleric Acid 95.0+%, TCI America™

Catalog No. p-7124942
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13721G 1 g
C13725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13721G Supplier TCI America Supplier No. C13721G

Chemical Identifiers

CAS 32653-34-2
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00191403
InChI Key QMYSXXQDOZTXAE-WHFBIAKZSA-N
Synonym (2S,3S)-2-Chloro-3-methylpentanoic Acid
PubChem CID 11051847
IUPAC Name (2S,3S)-2-chloro-3-methylpentanoic acid
SMILES CCC(C)C(C(=O)O)Cl

Specifications

Color Yellow
Boiling Point 125°C
Quantity 1 g
Formula Weight 150.60
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (2S,3S)-2-Chloro-3-methylvaleric Acid