missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S,3S)-(+)-2,3-Butanediol, 99%, Thermo Scientific™
Chemical Identifiers
| CAS | 19132-06-0 |
|---|---|
| Molecular Formula | C4H10O2 |
| Molecular Weight (g/mol) | 90.12 |
| MDL Number | MFCD00063648 |
| InChI Key | OWBTYPJTUOEWEK-IMJSIDKUSA-N |
| Synonym | 2s,3s-butane-2,3-diol, 2s,3s-+-2,3-butanediol, s,s-2,3-butanediol, s,s-+-2,3-butanediol, s,s-butane-2,3-diol, unii-7e9uxg71s1, 2,3-butanediol, +, s,s-2,3-butylene glycol, 2,3-butanediol, s-r*,r*, 2s,3s-2,3-butanediol |
| PubChem CID | 439888 |
| ChEBI | CHEBI:16812 |
| IUPAC Name | (2S,3S)-butane-2,3-diol |
| SMILES | CC(C(C)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC247735000
|
Thermo Scientific Chemicals
247735000 |
500mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC247730025
|
Thermo Scientific Chemicals
247730025 |
2.5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chiral derivatization reagentChemical Identifiers
| 19132-06-0 | |
| 90.12 | |
| OWBTYPJTUOEWEK-IMJSIDKUSA-N | |
| 439888 | |
| (2S,3S)-butane-2,3-diol |
| C4H10O2 | |
| MFCD00063648 | |
| 2s,3s-butane-2,3-diol, 2s,3s-+-2,3-butanediol, s,s-2,3-butanediol, s,s-+-2,3-butanediol, s,s-butane-2,3-diol, unii-7e9uxg71s1, 2,3-butanediol, +, s,s-2,3-butylene glycol, 2,3-butanediol, s-r*,r*, 2s,3s-2,3-butanediol | |
| CHEBI:16812 | |
| CC(C(C)O)O |
Specifications
| 19132-06-0 | |
| 100.0 | |
| 179.0°C to 182.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4309 to 1.4329 | |
| MFCD00063648 | |
| 01, 2183 | |
| 0.98 | |
| + 13.00 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and diisopropyl ether | |
| CC(C(C)O)O | |
| 90.12 | |
| CHEBI:16812 | |
| 99% |
| 98.5 | |
| 0.9800g/mL | |
| 85°C | |
| 98.5% min. (GC) | |
| C4H10O2 | |
| CH3CH(OH)CH(OH)CH3 | |
| 500mg | |
| 01,82 | |
| 15, 1571 | |
| 2s,3s-butane-2,3-diol, 2s,3s-+-2,3-butanediol, s,s-2,3-butanediol, s,s-+-2,3-butanediol, s,s-butane-2,3-diol, unii-7e9uxg71s1, 2,3-butanediol, +, s,s-2,3-butylene glycol, 2,3-butanediol, s-r*,r*, 2s,3s-2,3-butanediol | |
| OWBTYPJTUOEWEK-IMJSIDKUSA-N | |
| (2S,3S)-butane-2,3-diol | |
| 439888 | |
| 90.12 | |
| (2S, 3S)-(+)-2, 3-Butanediol, 99% |
Safety and Handling
AIR SENSITIVE
HYGROSCOPIC
TSCA : TSCA
RUO â Research Use Only