missing translation for 'onlineSavingsMsg'
Learn More

(2S,3R)-3-Phenylpyrrolidine-2-carboxylic acid, 98%

Catalog No. AC290081000
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC290081000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC290081000 Supplier Thermo Scientific Chemicals Supplier No. 290081000
Only null left

CAS: 118758-48-8 | C11H13NO2 | 191.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118758-48-8
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00142984
InChI Key VDEMEKSASUGYHM-QVDQXJPCSA-N
Synonym 3-phenyl-l-proline, 3-phenyl proline, 2s-3-phenylpyrrolidine-2-carboxylic acid
PubChem CID 133109024
IUPAC Name (2R)-3-phenylpyrrolidine-2-carboxylic acid
SMILES C1CNC(C1C2=CC=CC=C2)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 234°C to 236°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Specific Rotation Condition + 84.00 (25.00°C c=0.75,6 N HCl)
Specific Rotation + 84
Formula Weight 191.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (2S, 3R)-3-Phenylpyrrolidine-2-carboxylic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse ca

RUO – Research Use Only