missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2R,8aS)-(+)-(Camphorylsulfonyl)oxaziridine 95.0+%, TCI America™
Supplier: TCI America C13265G
Specifications
| (2R,8aS)-(+)-(Camphorylsulfonyl)oxaziridine [Asymmetric Oxidizing Reagent] | |
| 167°C | |
| C10H15NO3S | |
| 5 g | |
| GBBJBUGPGFNISJ-SKVWKECBSA-N | |
| 229.294 | |
| 229.29 | |
| Crystalline Powder |
| 104322-63-6 | |
| White-Yellow | |
| MFCD00075319 | |
| 1r---10-camphorsulfonyl oxaziridine | |
| CC1(C2CCC13CS(=O)(=O)N4C3(C2)O4)C | |
| 86278372 | |
| ≥95.0% (T) |
Chemical Identifiers
| 104322-63-6 | |
| 229.294 | |
| GBBJBUGPGFNISJ-SKVWKECBSA-N | |
| 86278372 |
| C10H15NO3S | |
| MFCD00075319 | |
| 1r---10-camphorsulfonyl oxaziridine | |
| CC1(C2CCC13CS(=O)(=O)N4C3(C2)O4)C |
Safety and Handling
TSCA : No