missing translation for 'onlineSavingsMsg'
Learn More

(2R,8aS)-(+)-(Camphorylsulfonyl)oxaziridine 95.0+%, TCI America™

Catalog No. C13261G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13261G 1 g
C13265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13261G Supplier TCI America Supplier No. C13261G

Chemical Identifiers

CAS 104322-63-6
Molecular Formula C10H15NO3S
Molecular Weight (g/mol) 229.294
MDL Number MFCD00075319
InChI Key GBBJBUGPGFNISJ-SKVWKECBSA-N
Synonym 1r---10-camphorsulfonyl oxaziridine
PubChem CID 86278372
SMILES CC1(C2CCC13CS(=O)(=O)N4C3(C2)O4)C

Specifications

Melting Point 167°C
Color White-Yellow
Quantity 1 g
Formula Weight 229.29
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (2R,8aS)-(+)-(Camphorylsulfonyl)oxaziridine [Asymmetric Oxidizing Reagent]