missing translation for 'onlineSavingsMsg'
Learn More

(2R,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%, Thermo Scientific™

Chemical Identifiers

CAS 98760-08-8
Molecular Formula C15H21NO3
Molecular Weight (g/mol) 263.34
MDL Number MFCD00671705,MFCD02258997
InChI Key NVPOUMXZERMIJK-UHFFFAOYNA-N
Synonym 2r,3s-3-tert-butoxycarbonyl amino-1,2-epoxy-4-phenylbutane, 2r,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, tert-butyl s-1-r-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl 1s,2r-oxiranyl-2-phenylethyl carbamate, threo-n-boc-l-phenylalanine epoxide, tert-butyl n-1s-1-2r-oxiran-2-yl-2-phenylethyl carbamate, 2r,3s-3-n-boc-amino-1-oxirane-4-phenylbutane, 2r,3s-3-tert-boc amino-1,2-epoxy-4-phenylbutane, atazanavir impurity c, pubchem5823
PubChem CID 9813904
IUPAC Name tert-butyl N-[(1S)-1-[(2R)-oxiran-2-yl]-2-phenylethyl]carbamate
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C1CO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC395010010
View Documents Promotion Details
N/A 1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC395010050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

98760-08-8
263.34
NVPOUMXZERMIJK-UHFFFAOYNA-N
9813904
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C1CO1
C15H21NO3
MFCD00671705,MFCD02258997
2r,3s-3-tert-butoxycarbonyl amino-1,2-epoxy-4-phenylbutane, 2r,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, tert-butyl s-1-r-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl 1s,2r-oxiranyl-2-phenylethyl carbamate, threo-n-boc-l-phenylalanine epoxide, tert-butyl n-1s-1-2r-oxiran-2-yl-2-phenylethyl carbamate, 2r,3s-3-n-boc-amino-1-oxirane-4-phenylbutane, 2r,3s-3-tert-boc amino-1,2-epoxy-4-phenylbutane, atazanavir impurity c, pubchem5823
tert-butyl N-[(1S)-1-[(2R)-oxiran-2-yl]-2-phenylethyl]carbamate

Specifications

98760-08-8
100.0
C15H21NO3
2r,3s-3-tert-butoxycarbonyl amino-1,2-epoxy-4-phenylbutane, 2r,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, tert-butyl s-1-r-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl 1s,2r-oxiranyl-2-phenylethyl carbamate, threo-n-boc-l-phenylalanine epoxide, tert-butyl n-1s-1-2r-oxiran-2-yl-2-phenylethyl carbamate, 2r,3s-3-n-boc-amino-1-oxirane-4-phenylbutane, 2r,3s-3-tert-boc amino-1,2-epoxy-4-phenylbutane, atazanavir impurity c, pubchem5823
NVPOUMXZERMIJK-UHFFFAOYNA-N
tert-butyl N-[(1S)-1-[(2R)-oxiran-2-yl]-2-phenylethyl]carbamate
9813904
98%
97.5
97.5% min. (HPLC)
MFCD00671705,MFCD02258997
Solubility in water: <0.1%
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C1CO1
263.34
263.34
(2R, 3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane
Videos
SDS
Documents

Documents

RUO – Research Use Only