missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2R,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 395010010
| Quantity | 1g |
|---|---|
| Packaging | Glass bottle |
Chemical Identifiers
| 98760-08-8 | |
| 263.34 | |
| NVPOUMXZERMIJK-UHFFFAOYNA-N | |
| 9813904 | |
| CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C1CO1 |
| C15H21NO3 | |
| MFCD00671705,MFCD02258997 | |
| 2r,3s-3-tert-butoxycarbonyl amino-1,2-epoxy-4-phenylbutane, 2r,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, tert-butyl s-1-r-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl 1s,2r-oxiranyl-2-phenylethyl carbamate, threo-n-boc-l-phenylalanine epoxide, tert-butyl n-1s-1-2r-oxiran-2-yl-2-phenylethyl carbamate, 2r,3s-3-n-boc-amino-1-oxirane-4-phenylbutane, 2r,3s-3-tert-boc amino-1,2-epoxy-4-phenylbutane, atazanavir impurity c, pubchem5823 | |
| tert-butyl N-[(1S)-1-[(2R)-oxiran-2-yl]-2-phenylethyl]carbamate |
Specifications
| (2R, 3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane | |
| 97.5 | |
| 97.5% min. (HPLC) | |
| C15H21NO3 | |
| 1g | |
| Solubility in water: <0.1% | |
| CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C1CO1 | |
| 263.34 | |
| 263.34 |
| 98760-08-8 | |
| 100.0 | |
| Glass bottle | |
| MFCD00671705,MFCD02258997 | |
| 2r,3s-3-tert-butoxycarbonyl amino-1,2-epoxy-4-phenylbutane, 2r,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, tert-butyl s-1-r-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl 1s,2r-oxiranyl-2-phenylethyl carbamate, threo-n-boc-l-phenylalanine epoxide, tert-butyl n-1s-1-2r-oxiran-2-yl-2-phenylethyl carbamate, 2r,3s-3-n-boc-amino-1-oxirane-4-phenylbutane, 2r,3s-3-tert-boc amino-1,2-epoxy-4-phenylbutane, atazanavir impurity c, pubchem5823 | |
| NVPOUMXZERMIJK-UHFFFAOYNA-N | |
| tert-butyl N-[(1S)-1-[(2R)-oxiran-2-yl]-2-phenylethyl]carbamate | |
| 9813904 | |
| 98% |