missing translation for 'onlineSavingsMsg'
Learn More

(2R,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid, 97%

CAS: 73397-28-1 | C12H23NO5 | 261.318 g/mol

Supplier:  Thermo Scientific Chemicals H5280603

Quantity 1 g

 View more versions of this product

Catalog No. AAH5280603


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

73397-28-1
261.318
DJZCWTDKDFJARG-DTWKUNHWSA-N
10377964
CC(C)CC(C(C(=O)O)O)NC(=O)OC(C)(C)C
C12H23NO5
MFCD04972275
boc-2r,3s-3-amino-2-hydroxy-5-methylhexanoicacid, 2r,3s-3-boc-amino-2-hydroxy-5-methylhexanoic acid, n-boc-2r,3s-2-hydroxy-3-amino-5-methylhexanoic acid, 2r,3s-3-tert-butoxycarbonyl amino-2-hydroxy-5-methylhexanoic acid, 2r,3s-2-hydroxy-3-tert-butoxycarbonylamino-5-methylhexanoic acid
(2R,3S)-2-hydroxy-5-methyl-3-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid

Specifications

(2R,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid
C12H23NO5
MFCD04972275
DJZCWTDKDFJARG-DTWKUNHWSA-N
(2R,3S)-2-hydroxy-5-methyl-3-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
10377964
97%
73397-28-1
1 g
boc-2r,3s-3-amino-2-hydroxy-5-methylhexanoicacid, 2r,3s-3-boc-amino-2-hydroxy-5-methylhexanoic acid, n-boc-2r,3s-2-hydroxy-3-amino-5-methylhexanoic acid, 2r,3s-3-tert-butoxycarbonyl amino-2-hydroxy-5-methylhexanoic acid, 2r,3s-2-hydroxy-3-tert-butoxycarbonylamino-5-methylhexanoic acid
CC(C)CC(C(C(=O)O)O)NC(=O)OC(C)(C)C
261.318
261.32
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions