Learn More
Sarstedt Inc 2ML SC MTUBE CBS GWB ST PK100 DFS Item
Supplier:  Sarstedt Inc 72694006
2ml Screw Cap Micro Tube Conical Bottom Skirted Graduations and Writing Block O-Ring Cap Assembled Sterile
Catalog No. 50809242PK
Only null left
Explore available promotions