missing translation for 'onlineSavingsMsg'
Learn More

2H-Perfluoro-5-methyl-3,6-dioxanonane, 97%, Thermo Scientific™

Catalog No. AAL1695406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1695406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1695406 Supplier Thermo Scientific Chemicals Supplier No. L1695406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 3330-14-1
Melting Point -122°C
Density 1.659
Boiling Point 103°C to 105°C
Flash Point None
Molecular Formula C8HF17O2
Refractive Index 1.3005
Quantity 5 g
MDL Number MFCD00054714
Synonym 2h-perfluoro-5-methyl-3,6-dioxanonane, 1,1,1,2,2,3,3-heptafluoro-3-1,1,1,2,3,3-hexafluoro-3-1,2,2,2-tetrafluoroethoxy propan-2-yl oxy propane, propane, 1-1-difluoro 1,2,2,2-tetrafluoroethoxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2,3,3,3-heptafluoro, cf3cfhocf2cf cf3 ocf2cf2cf3, 2h-perfluoro 5-methyl-3,6-dioxanonane, 1,1,1,2,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy-3-1,2,2,2-tetrafluoroethoxy propane, 1,1,1,2,4,4,5,7,7,8,8,9,9,9-tetradecafluoro-5-trifluoromethyl-3,6-dioxanonane
InChI Key PYSYKOPZHYNYSZ-UHFFFAOYSA-N
SMILES C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F
IUPAC Name 1,1,1,2,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)-3-(1,2,2,2-tetrafluoroethoxy)propane
Molecular Weight (g/mol) 452.067
PubChem CID 102982
Formula Weight 452.07
Percent Purity 97%
Chemical Name or Material 2H-Perfluoro-5-methyl-3,6-dioxanonane
Show More Show Less
TSCA Yes
Recommended Storage Ambient temperatures