missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Luer Lock insert and body connectors with reducers, injection site

Description
The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.
Features
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
- Constructed in an ISO 7 clean room under cGMP conditions
Applications
- Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
- Preparing and storing buffer, culture media, and process liquids
- Collecting chromatography feeds and fractions
- Harvesting, storing, and transporting bulk drug product and drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry's largest component library, with over 2,000 unique components that include:
- Fittings – straight, reducing, elbow, T-style, X-style, cross-style
- Connectors –quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
- Filters (used for bioburden reduction ) –sterilization grade filters, vent filters
- Tubing –thermoplastic elastomers, platinum-cured silicone
- Sample ports –septum-style ports, needle-free ports
- Clamps –sliders, pinch clamps, crimp bands
BPC port specifications:
- Line 1: Luer lock insert connection with 4.8 mm (3/16 in.) barb, cap, slide clamp, and 4.8 x 4.1 mm (3/16 x 5/32 in.) reducer. Tubing: TPE 374, length: 30 cm (12 in.), ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
- Line 2: Luer lock body connection with 0.56 cm (0.22 in.) barb, plug, slide clamp, and 0.48 x 0.41 cm (0.12 x 0.16 in.) reducer. Tubing: TPE 374, length: 30 cm (12 in.), ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
- Line 3: Tubing: EVA, injection port
Specifications
Specifications
| Capacity (Metric) | 1 L |
| Material | Polyethylene |
| Content And Storage | Store at room temperature. |
| Dimensions | 32 x 14.7 cm (12.6 x 5.8 in.) |
| Outer Packaging | One 4-mil. poly bag |
| Quantity | 50/Cs. |
| Sterilization Method | Irradiated |
| Shipping Condition | Room Temperature |
| Product Line | Labtainer |
| Port Detail | Luer Lock Body Connector and Injection Site |
| Show More |