missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Luer Lock insert and body connectors with reducers, injection site
GREENER_CHOICE

Catalog No. SS00029I
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SS00029I 100 mL
SS00030I 250 mL
SS00031I 500 mL
SS00032I 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SS00029I Supplier Thermo Scientific™ Supplier No. SS00029I

The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.

The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.

Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions

Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands

BPC port specifications:
Line 1: Luer lock insert connection with 4.8 mm (3/16 in.) barb, cap, slide clamp, and 4.8 x 4.1 mm (3/16 x 5/32 in.) reducer. Tubing: TPE 374, length: 30 cm (12 in.), ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Line 2: Luer lock body connection with 0.56 cm (0.22 in.) barb, plug, slide clamp, and 0.48 x 0.41 cm (0.12 x 0.16 in.) reducer. Tubing: TPE 374, length: 30 cm (12 in.), ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Line 3: Tubing: EVA, injection port

Specifications

Capacity (Metric) 100 mL
Material Polyethylene
Content And Storage Store at room temperature.
Dimensions 16 x 10 cm (6.3 x 3.94 in.)
Outer Packaging One 4-mil. poly bag
Quantity 50/Cs.
Sterilization Method Irradiated
Shipping Condition Room Temperature
Product Line Labtainer
Port Detail Luer Lock Body Connector and Injection Site
Film Description ASI 26/77
Tubing Style TPE and EVA
Show More Show Less