missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Luer Lock insert and body connectors, injection site

Description
The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.
Features
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
- Constructed in an ISO 7 clean room under cGMP conditions
Applications
- Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
- Preparing and storing buffer, culture media, and process liquids
- Collecting chromatography feeds and fractions
- Harvesting, storing, and transporting bulk drug product and drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry's largest component library, with over 2,000 unique components that include:
- Fittings – straight, reducing, elbow, T-style, X-style, cross-style
- Connectors – quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
- Filters (used for bioburden reduction ) – sterilization grade filters, vent filters
- Tubing – thermoplastic elastomers, platinum-cured silicone
- Sample ports – septum-style ports, needle-free ports
- Clamps – sliders, pinch clamps, crimp bands
BPC port specifications:
- Line 1: Luer lock insert connection with 4.8 mm (3/16 in.) barb, cap, and slide clamp. Tubing: EVA + TPE 374
- Line 2: Luer lock body connection with 4.8 mm (3/16 in.) barb, cap, and slide clamp. Tubing: EVA + TPE 374
- Line 3: Tubing: EVA, injection port
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Material | Polyethylene |
| Content And Storage | Store at room temperature. |
| Dimensions | 25.9 x 13.7 cm (10.2 x 5.4 in.) |
| Outer Packaging | One 4-mil. poly bag |
| Quantity | 50/Cs. |
| Sterilization Method | Irradiated |
| Shipping Condition | Room Temperature |
| Product Line | Labtainer |
| Port Detail | Luer Lock Body Connector and Injection Site |
| Show More |