missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 2D Bar-coded Cryogenic Vial Storage Box for 81 Vials

Catalog No. 07200964
Change view
Click to view available options
Holds:
81 (2 mL Vials)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
07200964 81 (2 mL Vials)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07200964 Supplier Corning™ Supplier No. 8673
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For storing cryogenic vials. Corning™ 2D Bar-coded Cryogenic Vial Storage Box is made of polycarbonate.

  • Reusable white polycarbonate box holds 81 cryogenic vials in a 9 x 9 array
  • Compatible with both internal and external threaded vials

Specifications

Type Cryo Storage Box
Holds 81 (2 mL Vials)
Tube Array 9 x 9
Weight (Metric) 12 kg