missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ 42 cal Arc Goggle Hood System

Catalog No. 170012007
Encompass
Change view
Click to view available options
Arc Flash Rating:
42 cal/cm²
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Arc Flash Rating
170012007 42 cal/cm²
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 170012007 Supplier National Safety Apparel™ Supplier No. KITHP27

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kit Includes: OPF/Para-Aramid balaclava and Medium energy PureView™ arc goggle with anti-fog inner lens

  • Lightweight alternative for 42 cal with 360 degree head protection
  • Provides increased flexibility with excellent breathability
  • Kit includes durable and comfortable balaclava and goggle with anti-fog inner lens
  • Goggles feature baffled top and bottom vents for increased airflow and comfortable elastic strap to secure goggles
  • Protection extends below neckline
  • Stretch material keeps head and neck comfortable
  • Nose covering
  • Made in the USA

Specifications

Gender Unisex
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Arc Flash Rating 42 cal/cm²
Certifications/Compliance CAT 3, NFPA 70 E, ASTM F2178, OSHA 1910.269, ANSI/ISEA Z87.1
Closure Type None
Product Type Goggles
Size Universal
Material Westex™ UltraSoft™, OPF Para-Aramid/OPF Blend
Color Olive Green
Seam Type Stitched
Suspender None
Show More Show Less