Learn More
Alkali Scientific 26' Cassette of 24mm Lab Tape, Clear with Black Print
Catalog No. 502388661
Encompass
Proper identification of scientific samples is often a key objective of the research laboratory. The process of writing on sample tubes/containers can be messy and unreliable. It can also be difficult to find the time to access a computer and design a layout for expensive ink jet labels. This clear tape with black print is ideal for label identification of any laboratory vessel for nearly any laboratory application. Standard label tapes can be autoclaved, boiled or even frozen to temperatures as low as -50C.
Catalog No. 502388661 Supplier Alkali Scientific Supplier No. L901024CK
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Proper identification of scientific samples is often a key objective of the research laboratory. The process of writing on sample tubes/containers can be messy and unreliable. It can also be difficult to find the time to access a computer and design a layout for expensive ink jet labels. This clear tape with black print is ideal for label identification of any laboratory vessel for nearly any laboratory application. Standard label tapes can be autoclaved, boiled or even frozen to temperatures as low as -50C.