Learn More
Alkali Scientific 26' Cassette of 12mm Lab Tape, Green with White Print
Catalog No. 502388862
Encompass
Proper identification of scientific samples is often a key objective of the research laboratory. The process of writing on sample tubes/containers can be messy and unreliable. It can also be difficult to find the time to access a computer and design a layout for expensive ink jet labels. This green tape with white print is ideal for label identification of any laboratory vessel for nearly any laboratory application. Standard label tapes can be autoclaved, boiled or even frozen to temperatures as low as -50C.
Catalog No. 502388862 Supplier Alkali Scientific Supplier No. L901012GW
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Proper identification of scientific samples is often a key objective of the research laboratory. The process of writing on sample tubes/containers can be messy and unreliable. It can also be difficult to find the time to access a computer and design a layout for expensive ink jet labels. This green tape with white print is ideal for label identification of any laboratory vessel for nearly any laboratory application. Standard label tapes can be autoclaved, boiled or even frozen to temperatures as low as -50C.