Learn More
Celltreat Scientific Products 25mL Serological Pipet, Paper/Plastic Wrapped, Sterile
Catalog No. NC1772270
Encompass
CELLTREAT Scientific Products 25mL Serological Pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. Printed indicator arrow for quickly locating negative graduations when using a pipet controller. Now available in double case quantities. Double case serological pipets offer a price savings. Save up to 65% on shipping costs. Reduce your order frequency and cardboard waste.
Catalog No. NC1772270 Supplier Celltreat Scientific Products Supplier No. 229026B
CELLTREAT Scientific Products 25mL Serological Pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. Printed indicator arrow for quickly locating negative graduations when using a pipet controller. Now available in double case quantities. Double case serological pipets offer a price savings. Save up to 65% on shipping costs. Reduce your order frequency and cardboard waste.