Learn More
Stemcell Technologies Inc DNase I, Size: 25 mg
Catalog No. NC1538504
Encompass
Deoxyribonuclease I (DNase I) is an endonuclease consisting of a single glycosylated polypeptide chain with two disulfide bonds. DNase is often included in tissue dissociation protocols to digest DNA that has leaked into the dissociation medium as a result of cell damage. DNase I preferentially cleaves phosphodiester linkages adjacent to pyrimidine nucleotides in both single- and double-stranded DNA, yielding polynucleotides with 5-phosphate and 3-hydroxyl groups (Bernardi et al.). DNase I has been used for the digestion of human tissues such as microglia (Klegeris & McGeer), cartilage (Dunham & Koch), colon (Fukushima & Fiocchi), epithelium (Fukushima & Fiocchi), liver (Vatakis et al.), lung (Fujino et al.), neural (Fuja et al.), and stem cells (Kusuma et al.).
Catalog No. NC1538504 Supplier Stemcell Technologies Inc Supplier No. STM07469
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Deoxyribonuclease I (DNase I) is an endonuclease consisting of a single glycosylated polypeptide chain with two disulfide bonds. DNase is often included in tissue dissociation protocols to digest DNA that has leaked into the dissociation medium as a result of cell damage. DNase I preferentially cleaves phosphodiester linkages adjacent to pyrimidine nucleotides in both single- and double-stranded DNA, yielding polynucleotides with 5-phosphate and 3-hydroxyl groups (Bernardi et al.). DNase I has been used for the digestion of human tissues such as microglia (Klegeris & McGeer), cartilage (Dunham & Koch), colon (Fukushima & Fiocchi), epithelium (Fukushima & Fiocchi), liver (Vatakis et al.), lung (Fujino et al.), neural (Fuja et al.), and stem cells (Kusuma et al.).