missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ 250ml Separating Funnel - Borosilicate Glass, Pear Shaped, With Stopper And Rota flow Stopcock

250ml Separating Funnel - Borosilicate Glass, Pear Shaped, with Stopper and Rota flow Stopcock

Supplier:  Eisco™ CH0484C

Catalog No. S27999


Only null left

Description

Description

  • 250 mL Capacity
  • Screw Type Rotaflow Stopcock
  • Made of 3.3 borosilicate glass
  • Socket Size 19/26
  • Plastic stopper included
Specifications

Specifications

250 mL
20.32 cm
Pear-shaped Separatory Funnel
Borosilicate Glass
19/26
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions