missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ 25 mm Pendulum Bobs

25 mm pendulum bobs are important tools while studying the physics of pendulum motion including tension force, gravity force, centripetal force, potential energy, kinetic energy, and more. Designed for use with full pendulum set up, not included.

$14.70 - $70.87

Specifications

Product Type Sphere
For Use With (Application) Pendulum Motion
View More Specs

Includes: bob wth threaded hook

Products 4
Catalog Number Mfr. No. Material Price Quantity  
Catalog Number Mfr. No. Material Price Quantity  
S41506
View Documents
Eisco™
PH0306E
Aluminum
Each for $14.70
Only null left
 
S41504
View Documents
Eisco™
PH0306A
Brass
Each for $22.10
Only null left
 
S41505
View Documents
Eisco™
PH0306B
Copper
Each for $35.93
Only null left
 
S41507
View Documents
Eisco™
PH0306F
Iron, Copper, Aluminum
Each for $70.87
Only null left
 
Description

Description

  • Spherical pendulum bobs with hook,
  • 25mm in diameter
  • Hooks are threaded and removable
  • Useful for a variety physics experiments
  • Works well with Eisco pendulum clamp S96186

Warranty and Services

1 year limited manufacturer warranty

Specifications

Specifications

Sphere
Pendulum Motion
Videos
SDS
Documents

Documents