missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 25 mL Sterile Serological Pipets
GREENER_CHOICE

Catalog No. 07000366
Change view
Click to view available options
Packaging Type:
Bulk
Individual Paper/Plastic Wrap
Individual Plastic Wrap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type
07000366 Individual Paper/Plastic Wrap
07000367 Individual Plastic Wrap
07000368 Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07000366 Supplier Greiner Bio-One Supplier No. 760180

Drip-free pipetting design with high optical clarity and maximum accuracy.

  • High-grade polystyrene
  • Clear, easily legible graduation
  • Increased volume from negative graduations
  • Colored vertical stripe for ease in reading the volume
  • With synthetic fibrous filter material for protection against

Specifications

Aspirator Plug Yes
Autoclavable Not Autoclavable
Material Polystyrene
Sterility Sterile
For Use With (Application) Cell Culture, Microbiology
Graduated Yes
Graduations 2/10
Graduation Type Negative Graduations
Includes Synthetic Fibrous Filter Material
Negative Graduations Yes
Quantity 200 Pk.
Packaging Type Individual Paper/Plastic Wrap
Volume (Metric) 25 mL
Type Serological Pipet
Disposable Single-use
Color Code Red
Show More Show Less