Learn More
Fisherbrand™ 25 gsm Disposable Isolation Gown
Offers basic protection from splashes and spills. Disposable spunbonded polypropylene isolation gown.
$282.30 - $282.30
Specifications
Product Type Isolation Gown
Gender Unisex
No. of Pockets None
Material Spunbond Polypropylene
Color Yellow
View More Specs
Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
12-893-005XXL
View Documents
Fisherbrand™
12893005XXL
2X-Large
Case of 100 Each for $282.30
Only null left
Description
Description
These disposable polypropylene gowns are used to protect the wearer from contact with potentially infectious liquid and solid material.
Specifications
Specifications
Isolation Gown
None
Yellow
Single-use
Elastic
25 gsm
Unisex
Spunbond Polypropylene
Neck and Waist Ties
Non-sterile
Long Sleeve
Not Autoclavable
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'