missing translation for 'onlineSavingsMsg'
Learn More
Learn More
National Safety Apparel™ 25 Cal 32 in. Arc Flash Coat
Coat provides the highest level of protection in work with electrical exposures and hazards
Supplier: National Safety Apparel™ C04UTUK253X32
Description
- Breathable FR fabric
- FR fabric will NOT melt, drip or ignite
- Woven FR wrist cuffs
- Generous fit for easy movement
- Front hook and loop closure
- Inherently flame resistant for FR Protection that will NOT wash off.
Specifications
| FR Hook and Loop | |
| Reusable | |
| Cotton, Nylon | |
| 3X-Large | |
| Stitched | |
| CAT 3, ASTM F1506, OSHA 1910.269, NFPA 70 E, CSA Z462 |
| Khaki | |
| Unisex | |
| Collared | |
| Flame | |
| 25 cal/cm² |