missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 25-bp DNA Ladder

Catalog No. 931343
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
931343 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 931343 Supplier Applied Biosystems™ Supplier No. 931343

Designed for the sizing of double-stranded DNA in the range of 25 to 2,500 bp

  • 25-bp DNA Ladder is designed for the sizing of double-stranded DNA in the range of 25 to 2,500 bp. It consists of 13 individual chromatography-purified DNA fragments and has reference bands at 2000, 150, and 25 bp for easy orientation.
  • 25-bp DNA Ladder is ideal for separation on 1-4% agarose gels.
  • Sharp, clear bands—chromatography purified fragments for consistent and reliable results
  • Designed with Applied Biosystems microarrays in mind—bands were designed to work with multiple Applied Biosystems microarray platforms including CytoScan, Axiom, and OncoScan platforms

Specifications

Content And Storage • 1 x 0.5 mL 25 bp DNA Ladder, store at -20°C
• 1 x 0.5 mL 6X Orange DNA Loading Dye, store at -20°C

Concentration 6X
Gel Type Agarose
Ready to Load No
Size Range 25 to 2500 bp
Gel Compatibility Agarose Gels, E-Gel™ 48, E-Gel™ 96, TBE Gels, TBE-Urea Gels
Quantity 50 μg
Shipping Condition Dry Ice
Product Type DNA Ladder

For Research Use Only. Not for use in diagnostic procedures.