missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 24-well No-Bottom Microplates
Ideal for work with nonpolar proteins and peptides. Greiner Bio-One 24 Well No-Bottom Microplate features chimney-well design to help prevent cross-contamination.
Supplier: Greiner Bio-One 66200006
Description
- Virgin polystyrene construction
- Medium binding surface
- Designed without bottom
- Features well diameter design that offers one-half the well surface area of standard plates
- Allows 50% sample volume reduction without m
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Standard | |
Clear | |
10 μL | |
Conical Bottom (V-well) | |
-20°C to +60°C |
4 x 6 | |
Polystyrene | |
24 | |
10 | |
83 x 58 x 11 mm |