Learn More
Chemglass Life Sciences Complete Kit: 24/40 Glass Adapter with All 6 Threaded Vial Adapters: 13-425, 15-425, 18-425, 20-400, 22-400, 24-400 DFS Item
Supplier:  Chemglass Inc CG131840
24/40 Glass Adapter with All 6 Threaded Vial Adapters: 13-425, 15-425, 18-425, 20-400, 22-400, 24-400. Adapters permit the connection of standard laboratory dram sample vials directly to the vapor tube or a bump trap of a rotary evaporator. Adapters have a top outer joint of the listed size and a 20-400 G.P.I. thread at the bottom. A PTFE-lined, polypropylene, double-threaded connector attaches the vial to the adapter.
Catalog No. 50-871-911
Only null left
Explore available promotions