Learn More
Celltreat Scientific Products 225mL Conical Bottom Centrifuge Tube, Sterile
Supplier: Celltreat Scientific Products 229465
CELLTREAT Scientific Products 225mL Centrifuge Tube. For applications requiring large volume centrifugation. Tubes include molded graduation lines. Polyethylene (HDPE) cap with integral plug seal provides a secure closure. To suport tube during centrifugation, use of a rotor cushion is required. Temperature range of -80C to 120C. Max force (RCF) g 7500. Every case is printed with the lot number for quality traceability. Packaged sterile, 6/re-sealable bag, 48/case.