Learn More
Chemglass Inc 22 Gauge Needle, 6"" Long Metal Hubbed
Supplier:  Chemglass Inc CG307542
Encompass
22 Gauge Needle, 6" Long Metal Hubbed. Needle for CG-3070 and CG-3074 syringes. All needles are made of #304 full hard stainless steel and have a 22 beveled point for septum penetration along with general use. Removable needles come complete with the PTFE ferrule necessary for assembly.
Numéro de catalogue. CG307542
Il en reste null