Learn More
Mattek Corp 20MM GLS BTM PETRI DISH PK/10
Supplier: Mattek Corporation P35G1520CSLEEVE
35 mm Dish, No. 1.5 Coverslip, 20 mm Glass Diameter, Uncoated. MatTek glass bottom dishes combine the convenience of standard size 35 mm disposable plastic petri dishes with the optical quality of glass, providing superior microscopic images. For most applications, No. 1.5 is the preferred coverslip thickness, especially for optimizing image quality with high numerical aperture objectives. All MatTek dishes are sterilized with gamma-irradiation.