Learn More
Foxx Life Sciences Round Carboy, HDPE, 20L, with 83mm (83B) Foxx VersaCap and Large Handles
GREENER_CHOICE
Catalog No. 501050108
Encompass
392-1001-OEM: Carboy, Round Industrial, 20L HDPE, VersaCap 83mm. Foxx Life Sciences' 20L Round Carboys with 83mm Cap are great for solvent waste and liquid management. Made from HDPE material.
Catalog No. 501050108 Supplier Foxx Life Sciences Supplier No. 3921001OEM
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
392-1001-OEM: Carboy, Round Industrial, 20L HDPE, VersaCap 83mm. Foxx Life Sciences' 20L Round Carboys with 83mm Cap are great for solvent waste and liquid management. Made from HDPE material.