Learn More
Acrobiosystems ACROBiosystems 200x DDM CHS buffer DFS Item
Supplier:  ACROBiosystems Inc DC115ML
Others;5ML;This 200x detergent solution contains 10%(w/v) DDM and 2% (w/v) CHS that is necessary for solubilization of membrane proteins. DDM, or n-Dodecyl -D-Maltoside [CAS: 69227-93-6], is a maltoside based non-ionic detergent with a hydrophilic maltose head and a hydrophobic long chain alkyl tail. CHS, or Cholesteryl Hemisuccinate Tris Salt [CAS: 102601-49-0], is an ionizable anionic detergent that stabilizes large unilamellar vesicles. DDM/CHS micelles provide a more membrane-like environment for solubilization of membrane proteins, and help to keep structural integrity and activity. This 200x detergent solution is sterile-filtered and contains no other preservatives.
Numéro de catalogue. 50-210-7488
Il en reste null
Découvrez plus d'offres spéciales