Chemglass Inc Bottle, 2000mL, Storage, 29/42 Outer Joint, Single Neck
Catalog No. CG112607
Bottle, 2000mL, Storage, 29/42 Outer Joint, Single Neck. Bottle has a standard taper outer joint. Supplied without stopper. Bottles are available amberized for protection of light sensitive materials.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Bottle, 2000mL, Storage, 29/42 Outer Joint, Single Neck. Bottle has a standard taper outer joint. Supplied without stopper. Bottles are available amberized for protection of light sensitive materials.