missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ 20% Neutral Buffered Formalin (NBF)

Catalog No. 5720
Change view
Click to view available options
Capacity:
5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
5720 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5720 Supplier Epredia™ Supplier No. 5720

Transport, store and retrieve pathology specimens in a safe and cost effective manner with 20% NBF in various configurations. 5-gallon size.

Epredia Neutral Buffered Formalin (NBF) is a premixed AFIP formulated product that reduces exposure to toxic fumes Phosphate buffers are used to stabilize the pH between 6.8 and 7.2 Stringent quality control ensures a specific formalin concentration Stable to freezing temperatures Concentrate reduces storage space One gallon of concentrate to four gallons of distilled or deionized water ensures consistent results with those of the premixed 10% NBF

  • Neutral Buffered Formalin 20%
  • Epredia Neutral Buffered Formalin (NBF) 20% is a premixed AFIP formulated product that reduces exposure to toxic fumes
  • Phosphate buffers are used to stabilize the pH between 6.8 and 7.2
  • Stringent quality control ensures a formalin concentration of 10%
  • Stable to freezing temperatures
  • Concentrate reduces storage space
  • One gallon of concentrate to four gallons of distilled or deionized water ensures consistent results with those of the premixed 10% NBF

Specifications

Capacity 5 gal.
For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Formalin
Solution Type Formalin
pH 6.8 to 7.2
Form Liquid

Non-sterile