missing translation for 'onlineSavingsMsg'
Learn More

Enespro™ Enespro™ 20 cal Ultrasoft Arc Flash Coverall

Catalog No. p-7220115
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17001621 Small
17001622 Medium
17001623 Large
17001624 X-Large
17001625 2X-Large
17001626 3X-Large
17001627 4X-Large
17001628 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17001621 Supplier Enespro™ Supplier No. C88UWSM32
Only null left

Durable and comfortable ultrasoft coveralls offer a unique blend of 88% cotton and 12% high-tenacity nylon for a smooth feel. The inherently FR material delivers exceptional performance and reliability.

  • Action back allows for full range of motion
  • All metal covered to ensure no contact with skin
  • Chest pockets with flaps and hidden snap closures
  • Comfortable sewn-in elastic waist for better fit
  • Convenient side pass-through pockets for access to pant pockets
  • Two front pockets with pass-through for access to pants pockets
  • FR fabric will not melt, drip or ignite

Specifications

Product Type Coveralls
Size Small
Gender Unisex
Material Cotton, Nylon
Color Navy
Disposable Reusable
Waist Style Elastic (Stitched-in)
Ankle Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Protection Class CAT 2
Resistance Flame
Arc Flash Rating 20 cal/cm²
Certifications/Compliance ASTM F1506, OSHA 1910.269, NFPA 70E, CSA Z462
Suspender None
For Use With (Application) Electrical Contractors, Manufacturing, Oil and Gas
Style Coveralls
Show More Show Less