Learn More
National Safety Apparel™ 20 Cal UltraSoft Arc Flash Bib Overalls
For enhanced protection Westex™ Ultrasoft is the ideal choice for superior comfort, improved safety, and above average durability.
$544.95 - $725.33
Specifications
Closure Type Buckle
Color Navy
Material Cotton, Nylon
Seam Type Stitched
Suspender Adjustable
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17001365
View Documents
National Safety Apparel™
C45UWSM32
Small
Each for $544.95
Only null left
17001366
View Documents
National Safety Apparel™
C45UWMD32
Medium
Each for $544.95
Only null left
17001367
View Documents
National Safety Apparel™
C45UWLG32
Large
Each for $544.95
Only null left
17001368
View Documents
National Safety Apparel™
C45UWXL32
X-Large
Each for $544.95
Only null left
17001369
View Documents
National Safety Apparel™
C45UW2X32
2X-Large
Each for $599.42
Only null left
17001370
View Documents
National Safety Apparel™
C45UW3X32
3X-Large
Each for $599.42
Only null left
17001371
View Documents
National Safety Apparel™
C45UW4X32
4X-Large
Each for $659.36
Only null left
17001372
View Documents
National Safety Apparel™
C45UW5X32
5X-Large
Each for $725.33
Only null left
Description
Description
  • Arc Rating: 20 cal/cm²
  • CAT 2
  • Fabric/Material: 13 oz. Westex™ UltraSoft™
  • Breathable FR fabric
  • FR fabric will not melt, drip or ignite
  • Inherently flame resistant protection will not wash/wear out
  • Quick disconnect and easy adjustable shoulder straps for quick entry
  • FR hook and loop calf closure for easy boot entry
  • FR hook and loop fly closure
  • Made in the USA
Specifications
Specifications
Buckle
Cotton, Nylon
Adjustable
20 cal/cm²
Flame
Reusable
Navy
Stitched
Open
Standard
CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70 E, CSA Z462