missing translation for 'onlineSavingsMsg'
Learn More

2-Vinylpyridine (stabilized with TBC) 97.0+%, TCI America™

Catalog No. V002425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V002425ML 25 mL
V0024500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V002425ML Supplier TCI America Supplier No. V002425ML

Chemical Identifiers

CAS 100-69-6
Molecular Formula C7H7N
Molecular Weight (g/mol) 105.14
MDL Number MFCD00006355
InChI Key KGIGUEBEKRSTEW-UHFFFAOYSA-N
Synonym 2-vinylpyridine, pyridine, 2-ethenyl, pyridine, 2-vinyl, pyridine, ethenyl, vinylpyridine, 2-vinyl pyridine, alpha-vinylpyridine, vinyl pyridine, unii-dt4uv4nnkx, ccris 5238
PubChem CID 7521
IUPAC Name 2-ethenylpyridine
SMILES C=CC1=CC=CC=N1

Specifications

Melting Point -50°C
Boiling Point 158°C
Quantity 25 mL
UN Number 3073
Formula Weight 105.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Vinylpyridine (stabilized with TBC)
EINECSNumber (5)-0716
RTECSNumber UU1040000
TSCA Yes
Recommended Storage Freezer