missing translation for 'onlineSavingsMsg'
Learn More

2-Vinylpyridine, 97%, stabilized

Catalog No. AC158002500
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158002500 250 mL Glass bottle
AC158000050 5 mL Glass bottle
AC158000010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC158002500 Supplier Thermo Scientific Chemicals Supplier No. 158002500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100-69-6 | C7H7N | 105.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-69-6
Molecular Formula C7H7N
Molecular Weight (g/mol) 105.14
MDL Number MFCD00006355
InChI Key KGIGUEBEKRSTEW-UHFFFAOYSA-N
Synonym 2-vinylpyridine, pyridine, 2-ethenyl, pyridine, 2-vinyl, pyridine, ethenyl, vinylpyridine, 2-vinyl pyridine, alpha-vinylpyridine, vinyl pyridine, unii-dt4uv4nnkx, ccris 5238
PubChem CID 7521
IUPAC Name 2-ethenylpyridine
SMILES C=CC1=CC=CC=N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -50°C
Color Brown
Density 0.9983g/mL
Boiling Point 159°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5490 to 1.551
Linear Formula (C5H4N)CH=CH2
Quantity 250 mL
Beilstein 20, 256
Specific Gravity 0.9983
Solubility Information Solubility in water: 2.5g/100 mL in water (20°C). Other solubilities: soluble in dilute acids,soluble in hydrocarbons,alcohol,ketones and,esters,soluble in methanol,ethanol,dioxane
Stabilizer 0.1% tert.-butylcatechol
Viscosity 1.2 mPa.s (20°C)
Formula Weight 105.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Vinylpyridine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vap
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
EINECSNumber 202-879-8
RTECSNumber UU1040000
TSCA TSCA

RUO – Research Use Only