missing translation for 'onlineSavingsMsg'
Learn More

2-Vinylnaphthalene, 97%, stabilized

Catalog No. p-211326
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC140900050 5 g Glass bottle
AC140900250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC140900050 Supplier Thermo Scientific Chemicals Supplier No. 140900050

CAS: 827-54-3 | C12H10 | 154.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 827-54-3
Molecular Formula C12H10
Molecular Weight (g/mol) 154.21
MDL Number MFCD00004125
InChI Key KXYAVSFOJVUIHT-UHFFFAOYSA-N
Synonym 2-vinylnaphthalene, naphthalene, 2-ethenyl, 2-vinyl-naphthalene, beta-vinylnaphthalene, poly 2-vinylnaphthalene, unii-hzd8li91n1, hzd8li91n1, polyvinylnaphthalene, 2-vinyl naphthalene, 2-ethenyinaphthalene
PubChem CID 13230
ChEBI CHEBI:51325
IUPAC Name 2-ethenylnaphthalene
SMILES C=CC1=CC=C2C=CC=CC2=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 63°C to 67°C
Color Beige to White or Yellow
Boiling Point 135°C (18.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C10H7CH=CH2
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,acetone,benzene
Stabilizer 0.5% max. p-tert.-butylcatechol
Formula Weight 154.21
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Vinylnaphthalene, 98%
Show More Show Less
EINECSNumber 212-573-6
TSCA TSCA

RUO – Research Use Only