missing translation for 'onlineSavingsMsg'
Learn More

2-Undecanone 98.0+%, TCI America™

Catalog No. p-7137835
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
U000625ML 25 mL
U0006500ML 500 mL
2 options

Catalog No. U000625ML

Supplier: TCI America U000625ML

Only null left

Chemical Identifiers

CAS 112-12-9
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.30
MDL Number MFCD00009583
InChI Key KYWIYKKSMDLRDC-UHFFFAOYSA-N
Synonym 2-undecanone, methyl nonyl ketone, 2-hendecanone, ketone, methyl nonyl, undecanone, nonyl methyl ketone, rue ketone, 2-oxoundecane, methyl n-nonyl ketone, methylnonylketone
PubChem CID 8163
ChEBI CHEBI:17700
IUPAC Name undecan-2-one
SMILES CCCCCCCCCC(C)=O

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 mL
Formula Weight 170.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Undecanone
EINECSNumber (2)-0542
RTECSNumber YQ2820000
TSCA Yes