missing translation for 'onlineSavingsMsg'
Learn More

2-Undecanone, 98%

Catalog No. p-7023773
Change view
Click to view available options
Quantity:
2500 g
500 g
Catalog No. Quantity
AAA1765936 500 g
AAA176590E 2500 g
2 options

Catalog No. AAA1765936

Supplier: Thermo Scientific Chemicals A1765936

Only null left

CAS: 112-12-9 | C11H22O | 170.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-12-9
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.30
MDL Number MFCD00009583
InChI Key KYWIYKKSMDLRDC-UHFFFAOYSA-N
Synonym 2-undecanone, methyl nonyl ketone, 2-hendecanone, ketone, methyl nonyl, undecanone, nonyl methyl ketone, rue ketone, 2-oxoundecane, methyl n-nonyl ketone, methylnonylketone
PubChem CID 8163
ChEBI CHEBI:17700
IUPAC Name undecan-2-one
SMILES CCCCCCCCCC(C)=O

Specifications

Melting Point 11°C to 13°C
Density 0.825
Boiling Point 228°C
Flash Point 89°C (192°F)
Refractive Index 1.43
Quantity 500 g
UN Number UN3082
Beilstein 1749573
Merck Index 14,6104
Formula Weight 170.3
Percent Purity 98%
Chemical Name or Material 2-Undecanone
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 203-937-5
RTECSNumber YQ2820000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only