missing translation for 'onlineSavingsMsg'
Learn More

2-Trimethylsilylthiophene 98.0+%, TCI America™

Catalog No. T17065G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17065G Supplier TCI America Supplier No. T17065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18245-28-8
Molecular Formula C7H12SSi
Molecular Weight (g/mol) 156.318
MDL Number MFCD00005415
InChI Key OANGLGSZPSFVDY-UHFFFAOYSA-N
Synonym 2-thienyltrimethylsilane, 2-trimethylsilylthiophene, trimethyl thiophen-2-yl silane, silane, trimethyl-2-thienyl, trimethyl-2-thienylsilane, 2-trimethylsilyl thiophene, trimethyl 2-thienyl silane, amtsi021, acmc-1c71q, thien-2-yl-trimethyl-silane
PubChem CID 140360
IUPAC Name trimethyl(thiophen-2-yl)silane
SMILES C[Si](C)(C)C1=CC=CS1

Specifications

Quantity 5 g
UN Number 1993
Formula Weight 156.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Trimethylsilylthiophene