missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilylethynyl)pyridine, 97%

Catalog No. AAL1125103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1125103 1 g
AAL1125106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1125103 Supplier Thermo Scientific Chemicals Supplier No. L1125103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86521-05-3 | C10H13NSi | 175.306 g/mol

2-(Trimethylsilylethynyl)pyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trimethylsilylethynyl)pyridine is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 86521-05-3
Molecular Formula C10H13NSi
Molecular Weight (g/mol) 175.306
MDL Number MFCD00066349
InChI Key WOFPTESETJKCBH-UHFFFAOYSA-N
Synonym 2-trimethylsilylethynyl pyridine, 2-trimethylsilyl ethynyl pyridine, trimethyl 2-pyridin-2-ylethynyl silane, 2-2-trimethylsilyl ethynyl pyridine, timtec-bb sbb009057, trimethylsilylethynylpyridine, trimethylsilylethynyl pyridin, acmc-1bl66, 2-trimethylsilylethynylpyridine
PubChem CID 522859
IUPAC Name trimethyl(2-pyridin-2-ylethynyl)silane
SMILES C[Si](C)(C)C#CC1=CC=CC=N1

Specifications

Melting Point 40°C to 43°C
Boiling Point 85°C to 86°C (1 mmHg)
Refractive Index 1.533
Quantity 1 g
Beilstein 4176916
Solubility Information Insoluble in water.
Formula Weight 175.31
Percent Purity 97%
Chemical Name or Material 2-(Trimethylsilylethynyl)pyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only