missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)pyridine 96.0+%, TCI America™

Catalog No. T20215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20215G 5 g
T20211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20215G Supplier TCI America Supplier No. T20215G
Only null left

Chemical Identifiers

CAS 13737-04-7
Molecular Formula C8H13NSi
Molecular Weight (g/mol) 151.284
MDL Number MFCD06797181
InChI Key IBLKFJNZUSUTPB-UHFFFAOYSA-N
Synonym 2-trimethylsilyl pyridine, pyridine, 2-trimethylsilyl, 2-pyridyltrimethylsilane, 2-trimethylsilylpyridine, 2-pyridyl trimethylsilane, trimethyl pyridin-2-yl silane, amtsi028, acmc-1cb48, trimethyl 2-pyridinyl silane
PubChem CID 139573
IUPAC Name trimethyl(pyridin-2-yl)silane
SMILES C[Si](C)(C)C1=CC=CC=N1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 151.28
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trimethylsilyl)pyridine