missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trimethylsilyl)ethyl 3-Nitro-1H-1,2,4-triazole-1-carboxylate 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 258.31 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-(Trimethylsilyl)ethyl 3-Nitro-1H-1,2,4-triazole-1-carboxylate |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |