missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylacetyl)thiophene, 98%

Catalog No. p-7044637
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0756006 5 g
AAL0756014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0756006 Supplier Thermo Scientific Chemicals Supplier No. L0756006

CAS: 20409-48-7 | C9H12OS | 168.254 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20409-48-7
Molecular Formula C9H12OS
Molecular Weight (g/mol) 168.254
MDL Number MFCD00051668
InChI Key PMUOKYKJIKZPIR-UHFFFAOYSA-N
Synonym 2-trimethylacetyl thiophene, 2,2-dimethyl-1-thiophen-2-yl propan-1-one, 2,2-dimethyl-1-2-thienyl propan-1-one, 2-pivaloylthiophene, t-butyl-2-thienylketone, acmc-1cb4m, 2-2,2-dimethylpropionyl thiophene, 2,2-dimethyl-1-thiophenyl-1-propanone, 2.2-dimethyl-1-thiophenyl-1-propanone
PubChem CID 2776976
IUPAC Name 2,2-dimethyl-1-thiophen-2-ylpropan-1-one
SMILES CC(C)(C)C(=O)C1=CC=CS1

Specifications

Density 1.07
Boiling Point 114°C to 116°C (14 mmHg)
Flash Point 99°C (210°F)
Refractive Index 1.535
Quantity 5 g
Beilstein 116243
Formula Weight 168.26
Percent Purity 98%
Chemical Name or Material 2-(Trimethylacetyl)thiophene
Hazard Category H302-H317
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only