missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyrimidine-5-carboxylic acid, 97%

Catalog No. p-7035867
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54088MD 250 mg
AAH5408803 1 g
AAH5408806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH54088MD Supplier Thermo Scientific Chemicals Supplier No. H54088MD

CAS: 306960-77-0 | C6H3F3N2O2 | 192.10 g/mol

2-(Trifluoromethyl)pyrimidine-5-carboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 306960-77-0
Molecular Formula C6H3F3N2O2
Molecular Weight (g/mol) 192.10
MDL Number MFCD09054851
InChI Key RGLZXVNOHDUMRJ-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyrimidine-5-carboxylic acid, 2-trifluoromethyl-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylic acid, 2-trifluoromethyl, 5-pyrimidinecarboxylic acid, 2-trifluoromethyl-9ci, pubchem13190, 2-trifluoromethylpyrimidine-5-carboxylicacid, 2-trifluoromethyl pyrimidine-5-carboxylicacid, 2-trifluoromethyl-5-pyrimidinecarboxilic acid, 2-trifluoromethyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-trifluoromethyl
PubChem CID 10583889
IUPAC Name 2-(trifluoromethyl)pyrimidine-5-carboxylic acid
SMILES OC(=O)C1=CN=C(N=C1)C(F)(F)F

Specifications

Melting Point 170°C to 175°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 192.1
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)pyrimidine-5-carboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only