missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyrimidine-4-carboxamide, 97%, Thermo Scientific Chemicals

Catalog No. p-7035096
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5280303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5280303 Supplier Thermo Scientific Chemicals Supplier No. H5280303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 914348-10-0 | Molecular Formula: C6H4F3N3O | Molecular Weight: 191.113 g/mol

2-(Trifluoromethyl)pyrimidine-4-carboxamide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)pyrimidine-4-carboxamide is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 914348-10-0
Molecular Formula C6H4F3N3O
Molecular Weight (g/mol) 191.113
MDL Number MFCD05662689
InChI Key LBNILUATVOCQRF-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyrimidine-4-carboxamide, 4-carbamoyl-2-trifluoromethyl pyrimidine, 2-trifluoromethyl-4-pyrimidinecarboxamide, 2-trifluoromethyl-pyrimidine-4-carboxylic acid amide, 2-trifluoromethylpyrimidine-4-carboxylic acid amide
PubChem CID 17750126
IUPAC Name 2-(trifluoromethyl)pyrimidine-4-carboxamide
SMILES C1=CN=C(N=C1C(=O)N)C(F)(F)F

Specifications

Quantity 1 g
Formula Weight 191.11
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)pyrimidine-4-carboxamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only