missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyridine 95.0+%, TCI America™

Catalog No. T26831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26831G 1 g
T26835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26831G Supplier TCI America Supplier No. T26831G

Chemical Identifiers

CAS 368-48-9
Molecular Formula C6H4F3N
Molecular Weight (g/mol) 147.10
MDL Number MFCD02183562
InChI Key ATRQECRSCHYSNP-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyridine, 2-trifluoromethyl-pyridine, trifluoromethylpyridine, 2-1,1,1-trifluoromethyl pyridine, pyridine, trifluoromethyl, alpha,alpha,alpha-trifluoro-2-picoline, 2-cf3-pyridine, trifluoromethyl pyridine
PubChem CID 136199
IUPAC Name 2-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC=CC=N1

Specifications

Color Yellow
Boiling Point 140°C
Quantity 1 g
UN Number 1993
Formula Weight 147.10
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethyl)pyridine